GNE-317 ≥98%

Synonyms: GNE 317GNE3175-(6-(3-methoxyoxetan-3-yl)-7-methyl-4-morpholinothieno[3,2-d]pyrimidin-2-yl)pyrimidin-2-amineGNE-317

B1716-25 B1716-5
76268-520EA 145.93 USD
76268-520 76268-522
GNE-317 ≥98%
GNE-317

GNE-317 is a potent and selective PI3K inhibitor with potential anticancer activity.

Formula: C₁₉H₂₂N₆O₃S
MW: 414,48 g/mol
MDL Number: MFCD28900677
CAS Number: 1394076-92-6
https://us.vwr.com/stibo/web/std.lang.all/99/90/24139990.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR