L-779450 ≥98%

Danger

Synonyms: L 779450L7794502-Chloro-5-(2-phenyl-4-(pyridin-4-yl)-1H-imidazol-5-yl)phenolL-7794502-chloro-5-(2-phenyl-5-(pyridin-4-yl)-1H-imidazol-4-yl)phenol

B1709-5 B1709-25
76268-494EA 531.21 USD
76268-494 76268-492
L-779450 ≥98%
L-779450

L-779450 is a potent, ATP-competitive Raf kinase inhibitor (IC50 =10 nM) that displays > 7, > 30 and > 70-fold selectivity over p38α, GSK3β and Lck respectively.

Formula: C₂₀H₁₄ClN₃O
MW: 347,8 g/mol
MDL Number: MFCD12828750
CAS Number: 303727-31-3
https://us.vwr.com/stibo/web/std.lang.all/53/92/24105392.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR