PH-797804 ≥98%

Supplier: BioVision

Synonyms: 3-[3-bromo-4-[(2,4-difluorophenyl)methoxy]-6-methyl-2-oxopyridin-1-yl]-N,4-dimethylbenzamidePH797804PH 7978043-(4-(2,4-difluorobenzyloxy)-3-bromo-6-methyl-2-oxopyridin-1(2H)-yl)-N,4-dimethylbenzamidePH-797804

AB235572-25MG AB235572-5MG
76268-486EA 980.59 USD
76268-486 76268-484
PH-797804 ≥98%
PH-797804

PH-797804 is a novel pyridinone inhibitor of p38α with IC50 of 26 nM; 4-fold more selective versus p38β.

Formula: C₂₂H₁₉BrF₂N₂O₃
MW: 477,3 g/mol
MDL Number: MFCD18251426
CAS Number: 586379-66-0
https://us.vwr.com/stibo/web/std.lang.all/53/90/24105390.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR