RO5126766 ≥98%

Synonyms: CH-51267663-[[2-[(Methylaminosulfonyl)amino]-3-fluoropyridin-4-yl]methyl]-4-methyl-7-[(pyrimidin-2-yl)oxy]-2H-1-benzopyran-2-one3-[[3-fluoro-2-(methylsulfamoylamino)pyridin-4-yl]methyl]-4-methyl-7-pyrimidin-2-yloxychromen-2-oneCH 51267663-[[2-[(Methylaminosulphonyl)amino]-3-fluoropyridin-4-yl]methyl]-4-methyl-7-[(pyrimidin-2-yl)oxy]-2H-1-benzopyran-2-oneRO-5126766CH5126766RO 51267663-[[3-fluoro-2-(methylsulphamoylamino)pyridin-4-yl]methyl]-4-methyl-7-pyrimidin-2-yloxychromen-2-one

B1705-25 B1705-5
76268-794EA 904.81 USD
76268-794 76268-792
RO5126766 ≥98%
RO5126766

Ro5126766 (CH5126766) is a potent and selective dual RAF/MEK inhibitor. For SK-MEL-28, SK-MEL-2, MIAPaCa-2, and SW480 cell lines, the IC50 is determined by WST-8 assay is 65, 28, 40, and 46 nM, respectively.

Formula: C₂₁H₁₈FN₅O₅S
MW: 471,46 g/mol
CAS Number: 946128-88-7
https://us.vwr.com/stibo/web/std.lang.all/53/88/24105388.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR