BMS-582949 ≥98%

Warning

Synonyms: BMS582949BMS-582949BMS 5829494-((5-(cyclopropylcarbamoyl)-2-methylphenyl)amino)-5-methyl-N-propylpyrrolo[2,1-f][1,2,4]triazine-6-carboxamidePS 540446PS-540446PS540446

B1704-1 B1704-5
76268-788EA 145.93 USD
76268-788 76268-790
BMS-582949 ≥98%
BMS-582949

BMS-582949 is a potent and selective P38 mitogen-activated protein kinase (P38 MAPK) inhibitor.

Formula: C₂₂H₂₆N₆O₂
MW: 406,48 g/mol
CAS Number: 623152-17-0
https://us.vwr.com/stibo/web/std.lang.all/53/87/24105387.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR