PD318088 ≥98%

Supplier: BioVision

Synonyms: PD-318088pd 318088PD3180885-bromo-N-(2,3-dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodoanilino)benzamidepd318088PD 318088

AB235566-5MG AB235566-1MG
76268-464EA 980.59 USD
76268-464 76268-462
PD318088 ≥98%
PD318088

PD318088 is an allosteric and non-ATP competitive MEK1/2 inhibitor.

Formula: C₁₆H₁₃BrF₃IN₂O₄
MW: 561,09 g/mol
MDL Number: MFCD18384973
CAS Number: 391210-00-7
https://us.vwr.com/stibo/web/std.lang.all/53/80/24105380.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR