JNK-IN-7 ≥98%

Synonyms: JNK-IN-7JNK-IN73-(4-(Dimethylamino)but-2-enamido)-N-(4-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)benzamideJNK IN 73-[[(E)-4-(dimethylamino)but-2-enoyl]amino]-N-[4-[(4-pyridin-3-ylpyrimidin-2-yl)amino]phenyl]benzamideJNKIN-7JNKIN7

B1695-1 B1695-5
76268-454EA 145.93 USD
76268-454 76268-456
JNK-IN-7 ≥98%
JNK-IN-7

JNK-IN-7 is a selective JNK inhibitor with IC50 values of 1.54 nM, 1.99 nM, 0.75 nM to JNK1, JNK2, JNK3, respectively. It also inhibits phosphorylation of c-Jun, which is a direct substrate of JNK kinase.

Formula: C₂₈H₂₇N₇O₂
MW: 493,56 g/mol
MDL Number: MFCD25976765
CAS Number: 1408064-71-0
https://us.vwr.com/stibo/web/std.lang.all/53/78/24105378.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR