BI-78D3 ≥98%

Synonyms: BI 78D34-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1H-1,2,4-triazol-5-oneBI-78D3BI78D34-(2,3-dihydro-1,4-benzodioxin-6-yl)-3-[(5-nitro-1,3-thiazol-2-yl)sulphanyl]-1H-1,2,4-triazol-5-one4-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-5-((5-nitrothiazol-2-yl)thio)-4H-1,2,4-triazol-3-ol4-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-3-((5-nitrothiazol-2-yl)thio)-1H-1,2,4-triazol-5(4H)-one

B1686-25 B1686-5
76270-282EA 513.7 USD
76270-282 76270-280
BI-78D3 ≥98%
BI-78D3

BI 78D3 is a competitive c-Jun N-terminal kinase (JNK) inhibitor with IC50 of 280 nM that displays > 100 fold selectivity over p38α and no activity at mTOR and PI-3K.

Formula: C₁₃H₉N₅O₅S₂
MW: 379,37 g/mol
MDL Number: MFCD01313575
CAS Number: 883065-90-5
https://us.vwr.com/stibo/web/std.lang.all/53/71/24105371.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR