VX-11e ≥98%

Synonyms: 4-[2-(2-chloro-4-fluoroanilino)-5-methylpyrimidin-4-yl]-N-[(1S)-1-(3-chlorophenyl)-2-hydroxyethyl]-1H-pyrrole-2-carboxamideVX-11e(S)-4-(2-((2-Chloro-4-fluorophenyl)amino)-5-methylpyrimidin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamideVX11eVX 11e

B1681-5 B1681-25
76270-262EA 525.38 USD
76270-262 76270-260
VX-11e ≥98%
VX-11e

VX-11e is a potent, selective, and orally bioavailable inhibitor of ERK (Extracellular Signal-Regulated Kinase); antitumor agent.

Formula: C₂₄H₂₀Cl₂FN₅O₂
MW: 500,35 g/mol
MDL Number: MFCD18433366
CAS Number: 896720-20-0
https://us.vwr.com/stibo/web/std.lang.all/53/66/24105366.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR