DEL-22379 ≥98%

Warning

Synonyms: N-(3-((5-Methoxy-1H-indol-3-yl)methylene)-2-oxoindolin-5-yl)-3-(piperidin-1-yl)propanamideN-[(3Z)-3-[(5-methoxy-1H-indol-3-yl)methylidene]-2-oxo-1H-indol-5-yl]-3-piperidin-1-ylpropanamideDEL-22379DEL 22379DEL22379

B1680-5 B1680-25
76270-258EA 636.28 USD
76270-258 76270-256
DEL-22379 ≥98%
DEL-22379

DEL-22379 is an ERK dimerization inhibitor with IC50 of 0.5 μM.

Formula: C₂₆H₂₈N₄O₃
MW: 444,53 g/mol
MDL Number: MFCD00950163
CAS Number: 181223-80-3
https://us.vwr.com/stibo/web/std.lang.all/53/65/24105365.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR