VRT752271 ≥98%

Danger

Synonyms: VRT-752271N-[(1S)-1-(3-chlorophenyl)-2-hydroxyethyl]-4-[5-chloro-2-(propan-2-ylamino)pyridin-4-yl]-1H-pyrrole-2-carboxamideUlixertinibVRT752271BVD-523(S)-4-(5-Chloro-2-(isopropylamino)pyridin-4-yl)-N-(1-(3-chlorophenyl)-2-hydroxyethyl)-1H-pyrrole-2-carboxamideBVD523BVD 523

B1679-25 B1679-5
76270-252EA 135.88 USD
76270-252 76270-254
VRT752271 ≥98%
VRT752271

VRT752271 is a reversible, ATP-competitive inhibitor of ERK1 and ERK2 kinases, with IC50 of <0.3 nM against ERK2.

Formula: C₂₁H₂₂Cl₂N₄O₂
MW: 433,33 g/mol
MDL Number: MFCD22628898
CAS Number: 869886-67-9
https://us.vwr.com/stibo/web/std.lang.all/53/64/24105364.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR