DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)

Synonyms: DNA sodium saltThymonucleic acid sodium salt

B1677-10 B1677-5 B1677-1 B1677-250
76270-120EA 426.14 USD
76270-120 76270-118 76270-116 76270-122
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from salmon testes)
DNA sodium salt (Deoxyribonucleic acid sodium salt)

Deoxyribonucleic acid (DNA) is a long double-helical molecule containing genetic information. DNA is a double-stranded (ds) molecule. Each strand is composed of an ordered combination of four nucleotides, each nucleotide consisting of a purine or pyrimidine base (adenine, guanine, thymine, or cytosine) and linked by phosphodiester bonds. Sperm cells from salmon testes are a good source for non-mammalian DNA. It is used as blocking agent to reduce the background in nucleic acid hybridization techniques.

MDL Number: MFCD00130922
CAS Number: 9007-49-2
https://us.vwr.com/stibo/web/std.lang.all/53/63/24105363.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR