CNX-1351 ≥98%

Synonyms: CNX1351CNX 13511-(4-((2-(1H-indazol-4-yl)-4-morpholinothieno[3,2-d]pyrimidin-6-yl)methyl)piperazin-1-yl)-6-methylhept-5-ene-1,4-dioneCNX-1351

B1673-25 B1673-5
76270-066EA 466.85 USD
76270-066 76270-064
CNX-1351 ≥98%
CNX-1351

CNX1351 is a potent and selective inhibitor of PI3Kα with IC50 value of 6.8 nM

Formula: C₃₀H₃₅N₇O₃S
MW: 573,71 g/mol
CAS Number: 1276105-89-5
https://us.vwr.com/stibo/web/std.lang.all/53/59/24105359.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR