Acalisib ≥98%

Synonyms: 6-fluoro-3-phenyl-2-[(1S)-1-(7H-purin-6-ylamino)ethyl]quinazolin-4-one(S)-2-(1-((9H-purin-6-yl)amino)ethyl)-6-fluoro-3-phenylquinazolin-4(3H)-oneGS 9820GS9820AcalisibGS-9820

B1666-5 B1666-25
76270-022EA 116.72 USD
76270-022 76270-024
Acalisib ≥98%
Acalisib

Acalisib (GS-9820) is a potent and selective inhibitor of PI3Kδ with IC50 value of 12.7 nM.

Formula: C₂₁H₁₆FN₇O
MW: 401,4 g/mol
MDL Number: MFCD28411423
CAS Number: 870281-34-8
https://us.vwr.com/stibo/web/std.lang.all/53/53/24105353.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR