AMG319 ≥98%

Synonyms: (S)-N-(1-(7-fluoro-2-(pyridin-2-yl)quinolin-3-yl)ethyl)-9H-purin-6-amineAMG 319N-[(1S)-1-(7-fluoro-2-pyridin-2-ylquinolin-3-yl)ethyl]-7H-purin-6-amineAMG319

B1663-25 B1663-5
76270-020EA 525.38 USD
76270-020 76270-018
AMG319 ≥98%
AMG319

AMG319 is a potent and selective PI3Kδ inhibitor with IC50 of 18 nM, also inhibits PI3Kγ with IC50 of 850 nM.

Formula: C₂₁H₁₆FN₇
MW: 385,4 g/mol
CAS Number: 1608125-21-8
https://us.vwr.com/stibo/web/std.lang.all/53/52/24105352.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR