AZD8835 ≥98%

Danger

Synonyms: AZD 88351-(4-(5-(5-amino-6-(5-(tert-butyl)-1,3,4-oxadiazol-2-yl)pyrazin-2-yl)-1-ethyl-1H-1,2,4-triazol-3-yl)piperidin-1-yl)-3-hydroxypropan-1-oneAZD8835AZD-8835

B1661-5 B1661-25
76269-730EA 116.72 USD
76269-730 76269-732
AZD8835 ≥98%
AZD8835

AZD8835 is a potent and selective inhibitor of PI3Kδ and PI3Kα with IC50 of 5.7 nM and 6.2 nM, with selectivity against PI3Kβ (IC50 431 nM) and PI3Kγ (IC50 90 nM).

Formula: C₂₂H₃₁N₉O₃
MW: 469,55 g/mol
CAS Number: 1620576-64-8
https://us.vwr.com/stibo/web/std.lang.all/53/51/24105351.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR