GDC-0084 ≥98%

Synonyms: 5-[8,9-dihydro-6,6-dimethyl-4-(4-morpholinyl)-6H-[1,4]oxazino[4,3-e]purin-2-yl]-2-pyrimidinamine5-(6,6-dimethyl-4-morpholin-4-yl-8,9-dihydropurino[8,9-c][1,4]oxazin-2-yl)pyrimidin-2-amineGDC-00845-(6,6-Dimethyl-4-morpholino-8,9-dihydro-6H-[1,4]oxazino[4,3-e]purin-2-yl)pyrimidin-2-amineGDC0084RG 7666RG7666RG-7666GDC 0084

B1656-25
76270-004EA 494.46 USD
76270-004
GDC-0084 ≥98%
GDC-0084

GDC-0084 is a potent, selective inhibitor of Class I PI3K and mTOR with good penetration across the blood-brain barrier.

Formula: C₁₈H₂₂N₈O₂
MW: 382,42 g/mol
MDL Number: MFCD30187522
CAS Number: 1382979-44-3
https://us.vwr.com/stibo/web/std.lang.all/53/46/24105346.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR