GDC-0326 ≥98%

Synonyms: GDC0326GDC 0326(S)-2-((2-(1-isopropyl-1H-1,2,4-triazol-5-yl)-5,6-dihydrobenzo[f]imidazo[1,2-d][1,4]oxazepin-9-yl)oxy)propenamideGDC-0326

B1655-5 B1655-25
76270-000EA 525.38 USD
76270-000 76269-998
GDC-0326 ≥98%
GDC-0326

GDC-0326 is a potent and selective inhibitor of α-Isoform of Phosphoinositide 3-Kinase.

Formula: C₁₉H₂₂N₆O₃
MW: 382,42 g/mol
CAS Number: 1282514-88-8
https://us.vwr.com/stibo/web/std.lang.all/53/44/24105344.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR