A66 ≥98%

Synonyms: A 66A66(S)-N1-(2-(tert-butyl)-4'-methyl-[4,5'-bithiazol]-2'-yl)pyrrolidine-1,2-dicarboxamideA-66(2S)-1-N-[5-(2-tert-butyl-1,3-thiazol-4-yl)-4-methyl-1,3-thiazol-2-yl]pyrrolidine-1,2-dicarboxamide

B1654-25 B1654-5
76269-898EA 274.36 USD
76269-898 76269-900
A66 ≥98%
A66

A66 is a highly specific and selective p110α inhibitor with IC50 of 32 nM.

Formula: C₁₇H₂₃N₅O₂S₂
MW: 393,53 g/mol
MDL Number: MFCD22378485
CAS Number: 1166227-08-2
https://us.vwr.com/stibo/web/std.lang.all/53/43/24105343.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR