CC-122 ≥98%

Synonyms: 2,6-Piperidinedione3-(5-amino-2-methyl-4-oxoquinazolin-3(4H)-yl)piperidine-2,6-dione3-(5-amino-2-methyl-4-oxo-3(4H)-quinazolinyl)-CC 122CC122CC-122Avadomide

B1650-25 B1650-5
76269-714EA 274.36 USD
76269-714 76269-716
CC-122 ≥98%
CC-122

CC-122 is a novel agent for DLBCL with antitumor and immunomodulatory activity.CC-122 binds CRBN and degrades Aiolos and Ikaros resulting in a mimicry of IFN signaling and apoptosis in DLBCL.

Formula: C₁₄H₁₄N₄O₃
MW: 286,29 g/mol
MDL Number: MFCD29919294
CAS Number: 1015474-32-4
https://us.vwr.com/stibo/web/std.lang.all/53/39/24105339.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR