H3B-6527 ≥98%

Synonyms: H3B 6527H3B6527N-{2-[(6-{[(2,6-Dichloro-3,5-dimethoxyphenyl)carbamoyl](methyl)amino}-4-pyrimidinyl)amino]-5-(4-ethyl-1-piperazinyl)phenyl}acrylamideN-(2-((6-(3-(2,6-Dichloro-3,5-dimethoxyphenyl)-1-methylureido)pyrimidin-4-yl)amino)-5-(4-ethylpiperazin-1-yl)phenyl)acrylamideH3B-6527

B1647-25 B1647-5
76269-886EA 246.97 USD
76269-886 76269-888
H3B-6527 ≥98%
H3B-6527

H3B-6527 is an orally bioavailable inhibitor of human fibroblast growth factor receptor 4 (FGFR4), with potential antineoplastic activity.

Formula: C₂₉H₃₄Cl₂N₈O₄
MW: 629,54 g/mol
MDL Number: MFCD30377210
CAS Number: 1702259-66-2
https://us.vwr.com/stibo/web/std.lang.all/53/34/24105334.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR