KN-92 phosphate ≥98%

Synonyms: KN-92 phosphateN-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-4-methoxybenzenesulfonamideKN-92phosphoric acidKN92 phosphateKN 92 phosphateN-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-4-methoxybenzenesulphonamideKN92

B1644-5 B1644-25
76269-880EA 367.76 USD
76269-880 76269-878
KN-92 phosphate ≥98%
KN-92 phosphate

KN-92 is an inactive derivative of KN-93. KN-93 is a selective inhibitor of Ca2+/calmodulin-dependent kinase II (CaMKII), competitively blocking CaM binding to the kinase (Ki = 370 nM).

Formula: C₂₄H₂₅ClN₂O₃S.H₃PO₄
MW: 554,98 g/mol
MDL Number: MFCD24369012
CAS Number: 1135280-28-2
https://us.vwr.com/stibo/web/std.lang.all/53/32/24105332.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR