KN-92 hydrochloride ≥98%

Synonyms: hydrochloride2-[N-(4-methoxybenzenesulphonyl)]amino-N-(4-chlorocinnamyl)-N-methylbenzylamineKN92 hydrochlorideN-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-4-methoxybenzenesulfonamide hydrochlorideN-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-4-methoxybenzenesulphonamide hydrochlorideN-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-4-methoxybenzenesulfonamide2-[N-(4-methoxybenzenesulfonyl)]amino-N-(4-chlorocinnamyl)-N-methylbenzylamineKN-92 hydrochloridemonohydrochlorideN-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-4-methoxybenzenesulphonamideKN 92 hydrochloride

B1643-25 B1643-5
76269-874EA 98.07 USD
76269-874 76269-876
KN-92 hydrochloride ≥98%
KN-92 hydrochloride

KN-92 is an inactive derivative of KN-93. KN-93 is a selective inhibitor of Ca2+/calmodulin-dependent kinase II (CaMKII), competitively blocking CaM binding to the kinase (Ki = 370 nM).

Formula: C₂₄H₂₆Cl₂N₂O₃S
MW: 493,45 g/mol
MDL Number: MFCD25372041
CAS Number: 1431698-47-3
https://us.vwr.com/stibo/web/std.lang.all/53/31/24105331.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR