SCH-58261 ≥98%
Synonyms:
2-(furan-2-yl)-7-phenethyl-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine
, SCH58261
, SCH-58261
, SCH 58261
B1638-5
B1638-25
76269-854EA
99.24
USD
76269-854
76269-856
SCH-58261 ≥98%
SCH-58261
SCH-58261 is the adenosine (A2A) receptor competitive antagonist. Displays 323-, 53- and 100-fold selectivity over A1, A2B and A3 receptors, respectively.
Formula:
C₁₈H₁₅N₇O MW: 345,36 g/mol |
MDL Number:
MFCD00933778 CAS Number: 160098-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...