SH-4-54 ≥98%

Warning

Synonyms: 4-[(4-cyclohexylphenyl)methyl-[2-[methyl-(2,3,4,5,6-pentafluorophenyl)sulphonylamino]acetyl]amino]benzoic acidSH-4-54SH-4544-[(4-cyclohexylphenyl)methyl-[2-[methyl-(2,3,4,5,6-pentafluorophenyl)sulfonylamino]acetyl]amino]benzoic acidSH 4 544-(N-(4-Cyclohexylbenzyl)-2-((2,3,4,5,6-pentafluoro-N-methylphenyl)sulfonamido)acetamido)benzoic acid4-(N-(4-Cyclohexylbenzyl)-2-((2,3,4,5,6-pentafluoro-N-methylphenyl)sulphonamido)acetamido)benzoic acidSH4-54SH454

B1637-25 B1637-5
76269-850EA 105.07 USD
76269-850 76269-852
SH-4-54 ≥98%
SH-4-54

SH-4-54 is a most potent, small molecule, non-phosphorylated STAT3 inhibitor, strongly binds to STAT3 protein (KD = 300 nM), is selective for STAT3 over STAT1.

Formula: C₂₉H₂₇F₅N₂O₅S
MW: 610,59 g/mol
MDL Number: MFCD28143917
CAS Number: 1456632-40-8
https://us.vwr.com/stibo/web/std.lang.all/53/24/24105324.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR