TGR5-Receptor-Agonist ≥98%

Synonyms: TGR5TGR5-Receptor-AgonistMDK-00245MDK00245MDK 002453-(2-Chlorophenyl)-N-(4-chlorophenyl)-N,5-dimethylisoxazole-4-carboxamide3-(2-chlorophenyl)-N-(4-chlorophenyl)-N,5-dimethyl-4-isoxazolecarboxamide

B1635-25 B1635-5
76269-842EA 116.75 USD
76269-842 76269-844
TGR5-Receptor-Agonist ≥98%
TGR5-Receptor-Agonist

TGR5 Receptor Agonist, a potent TGR5(GPCR19) agonist, showed improved potency in the U2-OS cell assay (pEC50 = 6.8) and in melanophore cells (pEC50 = 7.5).

Formula: C₁₈H₁₄Cl₂N₂O₂
MW: 361.22 g/mol
MDL Number: MFCD17169972
CAS Number: 1197300-24-5
https://us.vwr.com/stibo/web/std.lang.all/53/22/24105322.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR