WIKI-4 ≥98%

Supplier: BioVision

Synonyms: 1-[(1S)-1-(4-chloro-3-fluorophenyl)-2-hydroxyethyl]-4-[2-[(2-methylpyrazol-3-yl)amino]pyrimidin-4-yl]pyridin-2-oneWIKI4WIKI 4WIKI-42-(3-((4-(4-Methoxyphenyl)-5-(pyridin-4-yl)-4H-1,2,4-triazol-3-yl)thio)propyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione

AB147053-5MG AB147053-25MG
76269-798EA 191.5 USD
76269-798 76269-800
WIKI-4 ≥98%
WIKI-4

WIKI4 is a potent inhibitor of Wnt/β-catenin signaling (EC50 ~ 75 nM); inhibits auto-ADP-ribosylation of tankyrase 2 (TNKS2) (IC50 ~15 nM).

Formula: C₂₉H₂₃N₅O₃S
MW: 521,59 g/mol
MDL Number: MFCD05885187
CAS Number: 838818-26-1
https://us.vwr.com/stibo/web/std.lang.all/18/02/24101802.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR