GDC0994 ≥98%

Synonyms: GDC0994RavoxertinibGDC 09941-[(1S)-1-(4-chloro-3-fluorophenyl)-2-hydroxyethyl]-4-[2-[(2-methylpyrazol-3-yl)amino]pyrimidin-4-yl]pyridin-2-one(S)-1-(1-(4-Chloro-3-fluorophenyl)-2-hydroxyethyl)-4-(2-((1-methyl-1H-pyrazol-5-yl)amino)pyrimidin-4-yl)pyridin-2(1H)-oneRG 7842GDC-0994RG7842RG-7842

B1629-25 B1629-5
76269-794EA 163.46 USD
76269-794 76269-796
GDC0994 ≥98%
GDC0994

GDC-0994 is highly selective inhibitor of extracellular signal-regulated kinase (ERK), ERK1 and ERK2, with biochemical potency of 1.1 nM and 0.3 nM, respectively.

Formula: C₂₁H₁₈ClFN₆O₂
MW: 440,86 g/mol
MDL Number: MFCD28143918
CAS Number: 1453848-26-4
https://us.vwr.com/stibo/web/std.lang.all/18/01/24101801.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR