GSK180736A ≥98%

Synonyms: GSK 180736AGSK180736A4-(4-fluorophenyl)-N-(1H-indazol-5-yl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxamideGSK-180736A

B1628-25 B1628-5
76269-792EA 467 USD
76269-792 76269-790
GSK180736A ≥98%
GSK180736A

GSK180736A is a G protein-coupled receptor kinase 2 (GRK2) inhibitor with an IC50 of 0.77 μM.

Formula: C₁₉H₁₆FN₅O₂
MW: 365,36 g/mol
MDL Number: MFCD30533616
CAS Number: 817194-38-0
https://us.vwr.com/stibo/web/std.lang.all/18/00/24101800.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR