HS-173 ≥98%

Warning

Synonyms: ethyl 6-(5-(phenylsulphonamido)pyridin-3-yl)imidazo[1,2-a]pyridine-3-carboxylateHS 173ethyl 6-(5-(phenylsulfonamido)pyridin-3-yl)imidazo[1,2-a]pyridine-3-carboxylateHS-173HS173

B1627-25 B1627-5
76269-786EA 106.35 USD
76269-786 76269-788
HS-173 ≥98%
HS-173

HS-173 is a novel PI3K inhibitor, that is used for cancer treatment. HS-173 inhibited the PI3K signaling pathway, and showed anti-proliferative effects on cancer cells.

Formula: C₂₁H₁₈N₄O₄S
MW: 422,46 g/mol
MDL Number: MFCD28023584
CAS Number: 1276110-06-5
https://us.vwr.com/stibo/web/std.lang.all/17/99/24101799.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR