Monensin monosodium salt

Synonyms: Monensin, Sodium SaltMonensin sodium salt

B1623-5 B1623-1 B1623-500
76270-838EA 147.71 USD
76270-838 76270-836 76270-840
Monensin monosodium salt
Monensin monosodium salt

Sodium Monensin, isolated from Streptomyces cinnamonensis, is a well-known representative of naturally polyether ionophore antibiotics. It preferentially forms complexes with monovalent cations to enable transport across lipid membranes.

Formula: C₃₆H₆₁NaO₁₁
MW: 692.85 g/mol
MDL Number: MFCD00077826
CAS Number: 22373-78-0
https://us.vwr.com/stibo/web/std.lang.all/17/96/24101796.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR