Regorafenib monohydrate ≥99.5%
Synonyms:
BAY 73 4506 monohydrate
, Tyrosine kinase inhibitor
, Regorafenib hydrate
, BAY 73-4506 monohydrate
, hydrate
, Regorafenib monohydrate
, 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
, 4-(4-(3-(4-chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide hydrate
, BAY 734506 monohydrate
Regorafenib monohydrate is a multi-targeted inhibitor tyrosine kinase
Formula:
C₂₁H₁₇ClF₄N₄O₄ MW: 500,83 g/mol |
MDL Number:
MFCD17170366 CAS Number: 1019206-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...