Odanacatib ≥98% (by HPLC)

Danger

Synonyms: Cathepsin K Inhibitor(S)-N-(1-Cyanocyclopropyl)-4-fluoro-4-methyl-2-(((S)-2,2,2-trifluoro-1-(4'-(methylsulfonyl)-[1,1'-biphenyl]-4-yl)ethyl)amino)pentanamideMK0822MK-0822MK 0822(S)-N-(1-Cyanocyclopropyl)-4-fluoro-4-methyl-2-(((S)-2,2,2-trifluoro-1-(4'-(methylsulphonyl)-[1,1'-biphenyl]-4-yl)ethyl)amino)pentanamide

B1617-5 B1617-25
76270-822EA 642.12 USD
76270-822 76270-820
Odanacatib ≥98% (by HPLC)
Odanacatib

Odanacatib is a potent, selective, and neutral inhibitor of cathepsin K

Formula: C₂₅H₂₇F₄N₃O₃S
MW: 525,56 g/mol
MDL Number: MFCD11042419
CAS Number: 603139-19-1
https://us.vwr.com/stibo/web/std.lang.all/17/92/24101792.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR