Lonafarnib ≥98%

Synonyms: SCH 663364-(2-(4-(8-Chloro-3,10-dibromo-6,11-dihydro-5H-benzo(5,6)cyclohepta(1,2-b)pyridin-11-yl)-1-piperidinyl)-2-oxoethyl)-1-piperidinecarboxamideFTI inhibitorfarnesyltransferase inhibitor(R)-4-(2-(4-(3,10-dibromo-8-chloro-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-yl)piperidin-1-yl)-2-oxoethyl)piperidine-1-carboxamideSarasarSCH-66336SCH66336

B1616-5 B1616-25
76270-818EA 811.42 USD
76270-818 76270-816
Lonafarnib ≥98%
Lonafarnib

Lonafarnib (SCH 66336) is a selectively farnesyltransferase inhibitor for H-ras, K-ras-4B and N-ras with IC50 of 1.9 nM, 5.2 nM and 2.8 nM, respectively.

Formula: C₂₇H₃₁Br₂ClN₄O₂
MW: 638,82 g/mol
MDL Number: MFCD06795138
CAS Number: 193275-84-2
https://us.vwr.com/stibo/web/std.lang.all/17/91/24101791.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR