Apatinib mesylate ≥98% (by HPLC)

Synonyms: ApatinibYN968D1YN-968D1N-(4-(1-cyanocyclopentyl)phenyl)-2-((4-methylpyridin-3-yl)amino)nicotinamide methanesulphonateYN 968D1N-(4-(1-cyanocyclopentyl)phenyl)-2-((4-methylpyridin-3-yl)amino)nicotinamide methanesulfonate

B1608-5 B1608-25
76270-634EA 642.12 USD
76270-634 76270-632
Apatinib mesylate ≥98% (by HPLC)
Apatinib mesylate

Apatinib mesylate is an orally bioavailable, small-molecule receptor tyrosine kinase inhibitor

Formula: C₂₅H₂₇N₅O₄S
MW: 493,58 g/mol
MDL Number: MFCD22420814
CAS Number: 1218779-75-9
https://us.vwr.com/stibo/web/std.lang.all/17/83/24101783.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR