Abiraterone ≥98%

Synonyms: CB 7598CB7598 (3S,8R,9S,10R,13S,14S)-10,13-dimethyl-17-pyridin-3-yl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-olCYP17 inhibitorCB-7598

B1607-5 B1607-25
76270-628EA 75.89 USD
76270-628 76270-630
Abiraterone ≥98%
Abiraterone

Abiraterone (CB-7598) is a potent steroidal cytochrome P450 17alpha-hydroxylase-17,20-lyase (CYP17) inhibitor with an IC50 at 4 nM

Formula: C₂₄H₃₁NO
MW: 349,51 g/mol
MDL Number: MFCD00924100
CAS Number: 154229-19-3
https://us.vwr.com/stibo/web/std.lang.all/17/82/24101782.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR