MRT67307 ≥98.8% (by HPLC)

Synonyms: N-(3-((5-Cyclopropyl-2-((3-(morpholinomethyl)phenyl)amino)pyrimidin-4-yl)amino)propyl)cyclobutanecarboxamideN-[3-[[5-cyclopropyl-2-[3-(morpholin-4-ylmethyl)anilino]pyrimidin-4-yl]amino]propyl]cyclobutanecarboxamide

B1604-5 B1604-25
76270-622EA 788.06 USD
76270-622 76270-620
MRT67307 ≥98.8% (by HPLC)
MRT67307

MRT67307 is a dual inhibitor of the IKKe and TBK-1

Formula: C₂₆H₃₆N₆O₂
MW: 464,62 g/mol
MDL Number: MFCD19443643
CAS Number: 1190378-57-4
https://us.vwr.com/stibo/web/std.lang.all/17/80/24101780.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR