Mps1-IN-1 ≥97% (by HPLC)

Synonyms: 1-[3-methoxy-4-[[4-(2-propan-2-ylsulfonylanilino)-1H-pyrrolo[2,3-b]pyridin-6-yl]amino]phenyl]piperidin-4-olMps 1 Inhibitor1-[3-methoxy-4-[[4-(2-propan-2-ylsulphonylanilino)-1H-pyrrolo[2,3-b]pyridin-6-yl]amino]phenyl]piperidin-4-olMps1 IN 1

B1603-5 B1603-25
76270-618EA 846.44 USD
76270-618 76270-616
Mps1-IN-1 ≥97% (by HPLC)
Mps1-IN-1

Mps1-IN-1 is a highly potent and selectibe Mpsl inhibitor

Formula: C₂₈H₃₃N₅O₄S
MW: 535,66 g/mol
MDL Number: MFCD22124489
CAS Number: 1125593-20-5
https://us.vwr.com/stibo/web/std.lang.all/17/79/24101779.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR