DL-3-Hydroxy-3-methylglutaryl Coenzyme A trisodium salt ≥95%
Supplier: BioVision
Synonyms:
HMG CoA Trisodium
AB285393-50MG
AB285393-5MG
AB285393-25MG
76270-726EA
2129.71
USD
76270-726
76270-724
76270-722
DL-3-Hydroxy-3-methylglutaryl Coenzyme A trisodium salt ≥95%
DL-3-Hydroxy-3-methylglutaryl Coenzyme A trisodium salt
DL-3-Hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) is a useful substrate for the enzyme 3-hydroxyl-3-methyl-glutaryl coenzyme A (HMG-CoA) reductase. HMG-CoA is the key intermediate in the biosynthsis of terpenes, cholesterol, and ketone bodies.
Formula:
C₂₇H₄₂N₇Na₂O₂0P₃S MW: 955,62 g/mol |
CAS Number:
103476-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...