Bendamustine hydrochloride

Danger

Synonyms: 5-[Bis(2-chloroethyl)amino]-1-methylbenzimidazole-2-butyric acid hydrochlorideCytostasane HClEP 3114-(5-(Bis(2-chloroethyl)amino)-1-methyl-1H-benzo[d]imidazol-2-yl)butanoic acid hydrochlorideSDX-15 HCl

B1596-5 B1596-25
76270-704EA 437.81 USD
76270-704 76270-702
Bendamustine hydrochloride
Bendamustine hydrochloride

Bendamustine is a novel alkylating agent with a benzimidazole ring giving it purine analogue-like properties.  Like other alkylating agents, bendamustine is thought to act by modifying and cross linking purine bases in DNA, thus inhibiting DNA, RNA and protein synthesis and leading to cell death in rapidly dividing cells.  Bendamustine appears to have a special activity against B cell leukemias and lymphomas.

Formula: C₁₆H₂₁Cl₂N₃O₂·HCl
MW: 394.71 g/mol
MDL Number: MFCD16879055
CAS Number: 3543-75-7
https://us.vwr.com/stibo/web/std.lang.all/17/73/24101773.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR