GSK-961081 ≥98% (by HPLC)

Synonyms: (R)-1-(3-((2-chloro-4-(((2-hydroxy-2-(8-hydroxy-2-oxo-1,2-dihydroquinolin-5-yl)ethyl)amino)methyl)-5-methoxyphenyl)amino)-3-oxopropyl)piperidin-4-yl [1,1'-biphenyl]-2-ylcarbamateTD-5959Batefenterol

B1590-5 B1590-25
76270-680EA 811.42 USD
76270-680 76270-412
GSK-961081 ≥98% (by HPLC)
GSK-961081

GSK-961081 is a bi-functional molecule with both muscarinic antagonism and β2-agonism (MABA) properties. GSK-961081 displays high affinity for hM2 (Ki = 1.4 nM), hM3 muscarinic receptors (Ki = 1.3 nM) and hβ2-adrenoceptors (Ki = 3.7 nM).

Formula: C₄₀H₄₂ClN₅O₇
MW: 740,25 g/mol
CAS Number: 743461-65-6
https://us.vwr.com/stibo/web/std.lang.all/17/67/24101767.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR