BI-847325 ≥98% (by HPLC)

Synonyms: 3-(3-(((4-((Dimethylamino)methyl)phenyl)amino)(phenyl)methylene)-2-oxoindolin-6-yl)-N-ethylpropiolamide(Z)-3-(3-((4-((dimethylamino)methyl)phenylamino)(phenyl)methylene)-2-oxoindolin-6-yl)-N-ethylpropiolamide

B1589-5 B1589-25
76270-408EA 204.31 USD
76270-408 76270-410
BI-847325 ≥98% (by HPLC)
BI-847325

BI-847325 is an ATP-competitive dual inhibitor of MEK and Aurora kinases. Potent inhibition of MEK1/2 and Aurora A/B kinases by BI-847325 was demonstrated in enzymatic and cellular assays.

Formula: C₂₉H₂₈N₄O₂
MW: 464,57 g/mol
MDL Number: MFCD28978743
CAS Number: 1207293-36-4
https://us.vwr.com/stibo/web/std.lang.all/17/66/24101766.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR