Solamargine ≥95%

Synonyms: Solasodine-L-rhamnosido-D-glucoside (3b,22a,25R)-Spirosol-5-en-3-yl O-6-deoxy-a-L-mannopyranosyl-(1→2)-O-[6-deoxy-a-L-mannopyranosyl-(1→4)]-b-D-glucopyranoside

B1586-25 B1586-5
76270-396EA 145.93 USD
76270-396 76270-398
Solamargine ≥95%
Solamargine

Solamargine is a major steroidal alkaloid glycoside extracted from a traditional Chinese medicinal herb, Solanum nigrum L. (SNL). It has been shown to inhibit growth and induce apoptosis of various cancer cells. Solamargine triggers cellular necrosis selectively in different types of human melanoma cancer cells through extrinsic mitochondrial death pathway.

Formula: C₄₅H₇₃NO₁₅
MW: 868,07 g/mol
CAS Number: 20311-51-7
https://us.vwr.com/stibo/web/std.lang.all/17/63/24101763.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR