BRD7116 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: N,N'-(sulphonylbis(4,1-phenylene))bis(2,2,3,3-tetramethylcyclopropane-1-carboxamide)N,N'-(sulphonyldi-4,1-phenylene)bis[2,2,3,3-tetramethylcyclopropanecarboxamide]N,N'-(sulfonylbis(4,1-phenylene))bis(2,2,3,3-tetramethylcyclopropane-1-carboxamide)N,N'-(sulfonyldi-4,1-phenylene)bis[2,2,3,3-tetramethylcyclopropanecarboxamide]

AB286219-25MG AB286219-5MG
76270-648EA 360.07 USD
76270-648 76270-650
BRD7116 ≥98% (by HPLC)
BRD7116

BRD 7116 is a selective inhibitor of leukemia stem cell (LSC) activity (EC₅₀ value of 200 nM). BRD 7116 does not affect normal hematopoietic stem cells or AML cell lines (EC₅₀s = 20 µM).

Formula: C₂₈H₃₆N₂O₄S
MW: 496,66 g/mol
MDL Number: MFCD02168947
CAS Number: 329059-55-4
https://us.vwr.com/stibo/web/std.lang.all/17/55/24101755.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR