GSK-2194069 ≥98% (by HPLC)

Synonyms: 4-[4-(benzofuran-5-yl)phenyl]-3-[[(3S)-1-(cyclopropanecarbonyl)pyrrolidin-3-yl]methyl]-1H-1,2,4-triazol-5-one

B1571-25 B1571-5
76270-590EA 811.42 USD
76270-590 76270-588
GSK-2194069 ≥98% (by HPLC)
GSK-2194069

GSK-2194069 is a potent and specific inhibitor of the β-ketoacyl reductase (KR) activity of hFAS (human Fatty Acid Synthase).

Formula: C₂₅H₂₄N₄O₃
MW: 428,48 g/mol
CAS Number: 1332331-21-1
https://us.vwr.com/stibo/web/std.lang.all/17/48/24101748.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR