BAY-1436032 ≥98% (by HPLC)

Danger

Synonyms: 3-(2-((4-(trifluoromethoxy)phenyl)amino)-1-((1R,5R)-3,3,5-trimethylcyclohexyl)-1H-benzo[d]imidazol-5-yl)propanoic acid

B1570-5 B1570-25
76270-586EA 758.88 USD
76270-586 76270-584
BAY-1436032 ≥98% (by HPLC)
BAY-1436032

BAY-1436032 is a potent and highly selective inhibitor of mutant Isocitrate Dehydrogenase 1 (mIDH1). BAY 1436032 strongly reduces 2-HG (2-hydroxyglutarate) levels in cells carrying IDH1-R132H, -R132C, -R132G, -R132S and -R132L mutations.

Formula: C₂₆H₃₀F₃N₃O₃
MW: 489,5 g/mol
CAS Number: 1803274-65-8
https://us.vwr.com/stibo/web/std.lang.all/17/47/24101747.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR