KRIBB11 ≥98% (by HPLC)

Supplier: BioVision

Synonyms: N2-(1H-Indazol-5-yl)-N6-methyl-3-nitropyridine-2,6-diamineN2-(1H-indazole-5-yl)-N6-methyl-3-nitropyridine-2,6-diamine

AB254320-25MG AB254320-5MG
76270-572EA 482.65 USD
76270-572 76270-574
KRIBB11 ≥98% (by HPLC)
KRIBB11

KRIBB11 is a Heat shock factor (HSF) inhibitor (IC₅₀ = 1.2 μM). KRIBB11 blocks the induction of HSF1 downstream target proteins such as HSP27 and HSP70. KRIBB11 induces growth arrest and apoptosis of HCT-116 cells.

Formula: C₁₃H₁₂N₆O₂
MW: 284,28 g/mol
MDL Number: MFCD30182306
CAS Number: 342639-96-7
https://us.vwr.com/stibo/web/std.lang.all/17/44/24101744.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR