Trovafloxacin mesylate ≥98% (by HPLC)

Synonyms: 7-[(1α,5α,6α)-6-Amino-3-azabicyclo[3.1.0]hex-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-1,4-dihydro-4-oxo-1,8-naphthyridine-3-carboxylic acid mesylateCP 99219

B1565-25 B1565-5
76270-570EA 788.06 USD
76270-570 76270-568
Trovafloxacin mesylate ≥98% (by HPLC)
Trovafloxacin mesylate

Trovafloxacin is a broad-spectrum fluoroquinolone antibiotic that inhibits the uncoiling of supercoiled DNA in bacteria by blocking the activity of topoisomerase IV (IC₅₀ = 3.02 µg/ml) and DNA gyrase (IC₅₀ = 7.13 µg/ml). Displays potent activity against gram-positive and gram-negative bacteria.

Formula: C₂₀H₁₅F₃N₄O₃.CH₃SO₃H
MW: 512,46 g/mol
MDL Number: MFCD00913361
CAS Number: 147059-75-4
https://us.vwr.com/stibo/web/std.lang.all/17/43/24101743.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR