    Sort by:
  • Search Within Results
Plinabulin
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Plinabulin (NPI-2358) is a vascular disrupting agent (VDA). Plinabulin binds to the colchicine binding site of β-tubulin preventing polymerization and...