    Sort by:
  • Search Within Results
Cyclic-di-GMP
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Cyclic di-GMP (c-di-GMP) is one of the most important and common bacterial second messenger. It is involved in numerous prokaryotic processes, includi...